Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

693553

Sigma-Aldrich

1,3-Bis(2,6-diisopropylphenyl)-4,5-dihydroimidazolium tetrafluoroborate

95%

Synonym(s):

4,5-Dihydro-1,3-bis(2,6-diisopropylphenyl)imidazolium tetrafluoroborate, N,N′-Bis(2,6-diisopropylphenyl)dihydroimidazolium tetrafluoroborate, SIPr-HBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39BF4N2
CAS Number:
Molecular Weight:
478.42
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

reagent type: catalyst

mp

>300

SMILES string

F[B-](F)(F)F.CC(C)c1cccc(C(C)C)c1N2CC[N+](=C2)c3c(cccc3C(C)C)C(C)C

InChI

1S/C27H39N2.BF4/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;2-1(3,4)5/h9-14,17-21H,15-16H2,1-8H3;/q+1;-1

InChI key

KFZBJQUHHALFSR-UHFFFAOYSA-N

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service