Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

574066

Sigma-Aldrich

1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride

Synonym(s):

1,3-Bis(mesityl)imidazolium chloride, 1,3-Dihydro-1,3-dimesityl-2H-imidazol-2-ylidene monohydrochloride, 1,3-Dimesitylimidazolium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25ClN2
CAS Number:
Molecular Weight:
340.89
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reagent type: ligand

mp

>300 °C (lit.)

SMILES string

[Cl-].Cc1cc(C)c(c(C)c1)-n2cc[n+](c2)-c3c(C)cc(C)cc3C

InChI

1S/C21H25N2.ClH/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;/h7-13H,1-6H3;1H/q+1;/p-1

InChI key

OTOSIXGMLYKKOW-UHFFFAOYSA-M

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thi Kim Hoang Trinh et al.
Physical chemistry chemical physics : PCCP, 21(31), 17036-17046 (2019-07-30)
Although N-heterocyclic carbenes (NHCs) have brought profound changes in catalytic organic synthesis, their generation generally requires an inert atmosphere and harsh conditions. To overcome these limitations, an air-stable NHC photogenerator has been developed involving two mild components: 1,3-bis(mesityl)imidazolium tetraphenylborate (IMesH+BPh4-)
Thi Kim Hoang Trinh et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(39), 9242-9252 (2019-04-26)
In the search of smarter routes to control the conditions of N-heterocyclic carbene (NHCs) formation, a two-component air-stable NHC photogenerating system is reported. It relies on the irradiation at 365 nm of a mixture of 2-isopropylthioxanthone (ITX) with 1,3-bis(mesityl)imidazoli(ni)um tetraphenylborate. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service