Skip to Content
MilliporeSigma
All Photos(1)

Documents

689556

Sigma-Aldrich

O-(2-Carboxyethyl)-O′-methyl-undecaethylene glycol

≥95% (oligomer purity)

Synonym(s):

mPEG acid (n=11)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H52O14
Molecular Weight:
588.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (oligomer purity)

form

chunks

SMILES string

COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(O)=O

InChI

1S/C26H52O14/c1-29-4-5-31-8-9-33-12-13-35-16-17-37-20-21-39-24-25-40-23-22-38-19-18-36-15-14-34-11-10-32-7-6-30-3-2-26(27)28/h2-25H2,1H3,(H,27,28)

InChI key

JMAKFNFKUHXFBH-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service