Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

680982

Sigma-Aldrich

Cyclopropylzinc bromide solution

0.5 M in THF

Synonym(s):

Bromocyclopropylzinc, Cyclopropane, zinc complex, Cyclopropylzinc(II) bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5BrZn
CAS Number:
Molecular Weight:
186.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

0.5 M in THF

density

0.969 g/mL at 25 °C
0.969 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Br[Zn]C1CC1
Br[Zn]C1CC1

InChI

1S/C3H5.BrH.Zn/c1-2-3-1;;/h1H,2-3H2;1H;/q;;+1/p-1
1S/C3H5.BrH.Zn/c1-2-3-1;;/h1H,2-3H2;1H;/q;;+1/p-1

InChI key

DUXRKEPSNWXJGV-UHFFFAOYSA-M
DUXRKEPSNWXJGV-UHFFFAOYSA-M

Application

Active nucleophile in Pd-catalyzed cross-coupling of heteroaryl halides.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Campbell, J. B. et al.
Synthetic Communications, 19, 2265-2265 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service