Skip to Content
MilliporeSigma
All Photos(1)

Documents

497517

Sigma-Aldrich

Benzylzinc bromide solution

0.5 M in THF

Synonym(s):

Benzylbromozinc, Benzylzinc(II) bromide, Bromo(phenylmethyl)zinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2ZnBr
CAS Number:
Molecular Weight:
236.42
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in THF

density

0.978 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Br[Zn]Cc1ccccc1

InChI

1S/C7H7.BrH.Zn/c1-7-5-3-2-4-6-7;;/h2-6H,1H2;1H;/q;;+1/p-1

InChI key

OQPRTUBCROVGHA-UHFFFAOYSA-M

Application

Benzylzinc bromide solution can be used as a nucleophile for the Mannich-type synthesis of 1,2-diarylethylpiperazines. It can also undergo C−C cross coupling reactions with aryl halides.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nickel Complexes Modified by O, N, O?-Ligands as Synthons for the Straightforward Synthesis of Highly Efficient Precatalysts for C-C Bond Formation.
Someya CI, et al.
Asian Journal of Organic Chemistry, 1(4), 322-326 (2012)
Straightforward three-component synthesis of diarylmethylpiperazines and 1, 2-diarylethylpiperazines.
Sengmany S, et al.
Tetrahedron, 63(18), 3672-3681 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service