Skip to Content
MilliporeSigma
All Photos(1)

Documents

498963

Sigma-Aldrich

Isobutylzinc bromide solution

0.5 M in THF

Synonym(s):

(2-Methylpropyl)zinc bromide, Bromo(2-methylpropyl)zinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2ZnBr
CAS Number:
Molecular Weight:
202.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

concentration

0.5 M in THF

density

0.943 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(C)C[Zn]Br

InChI

1S/C4H9.BrH.Zn/c1-4(2)3;;/h4H,1H2,2-3H3;1H;/q;;+1/p-1

InChI key

WYBHVVXVUCUWSE-UHFFFAOYSA-M

Application

Isobutylzinc bromide is an organozinc compound, which can be used as a reagent in the palladium-catalyzed Negishi cross-coupling reaction to construct a carbon-carbon bond by reacting with aryl or heteroaryl halides . It is also used to prepare regioselective unsaturated ketones by reacting with tert-butyl isocyanide via an allylic carbonylative coupling reaction in the presence of a nickel catalyst.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-Catalyzed Secondary Benzylic Imidoylative Reactions
Wang C, et al.
Organic Letters (2020)
Annexin-V stabilizes membrane defects by inducing lipid phase transition
Lin YC, et al.
Nature Communications, 11(1), 1-13 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service