Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

672564

Sigma-Aldrich

2-(Di-tert-butylphosphino)-1-(2-methoxyphenyl)-1H-pyrrole

95%

Synonym(s):

cataCXium® POMetB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28NOP
CAS Number:
Molecular Weight:
317.41
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

reaction suitability

reaction type: Asymmetric synthesis
reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

functional group

phosphine

SMILES string

COc1ccccc1-n2cccc2P(C(C)(C)C)C(C)(C)C

InChI

1S/C19H28NOP/c1-18(2,3)22(19(4,5)6)17-13-10-14-20(17)15-11-8-9-12-16(15)21-7/h8-14H,1-7H3

InChI key

JALZQSOGOMZLEK-UHFFFAOYSA-N

General description

sold in collaboration with Solvias AG

Legal Information

WO 2004/101581
cataCXium is a registered trademark of Umicore AG & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

cataCXium® ligands facilitate cross-coupling reactions, essential in polymer science, fine chemicals, and pharmaceutical industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service