Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

670855

Sigma-Aldrich

5-Bromo-1-pentanol

technical, ≥80% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11BrO
CAS Number:
Molecular Weight:
167.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥80% (GC)

refractive index

n20/D 1.482

functional group

bromo
hydroxyl

storage temp.

2-8°C

SMILES string

OCCCCCBr

InChI

1S/C5H11BrO/c6-4-2-1-3-5-7/h7H,1-5H2

InChI key

WJVQJXVMLRGNGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 - 122.0 °F

Flash Point(C)

40 - 50 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tian Hua et al.
Cell, 180(4), 655-665 (2020-02-01)
Human endocannabinoid systems modulate multiple physiological processes mainly through the activation of cannabinoid receptors CB1 and CB2. Their high sequence similarity, low agonist selectivity, and lack of activation and G protein-coupling knowledge have hindered the development of therapeutic applications. Importantly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service