Skip to Content
MilliporeSigma
All Photos(3)

Documents

B65586

Sigma-Aldrich

2-Bromoethanol

95%

Synonym(s):

Ethylene bromohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2OH
CAS Number:
Molecular Weight:
124.96
Beilstein:
878140
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.3 (vs air)

vapor pressure

2.4 mmHg ( 20 °C)

Assay

95%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

56-57 °C/20 mmHg (lit.)

density

1.763 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCCBr

InChI

1S/C2H5BrO/c3-1-2-4/h4H,1-2H2

InChI key

LDLCZOVUSADOIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromoethanol can be used for the preparation of 2-bromoethyl glycosides from acetylated sugars. It can also be used as a starting material to synthesize 2-bromoethyl methoxymethyl ether.
2-Bromoethanol is used in selective reduction of nitroarenes (PcFe(II)/NaBH4/2-bromoethanol catalyst system).

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Regio?and Stereoselective Intramolecular Hydrosilylation of α?Hydroxy Enol Ethers: 2, 3?syn?2?Methoxymethoxy?1, 3?Nonanediol: 1, 3?Nonanediol, 2?(methoxymethoxy)?,(R, R)?(?)?.
Tamao K, et al.
Organic Syntheses, 73, 94-94 (2003)
2-Bromoethyl glycosides: synthesis and characterisation.
Dahmen J, et al.
Carbohydrate Research, 116(2), 303-307 (1983)
S Khan et al.
Biochemical pharmacology, 45(2), 439-447 (1993-01-26)
The cytotoxicity of dibromoalkanes to isolated hepatocytes was proportional to the dibromoalkane concentration and increasing chain length of the dibromoalkane (C2-C6). The rapid hepatocyte glutathione (GSH) depletion which occurred upon addition of the dibromoalkanes was also dependent on the concentration
W Van Rillaer et al.
Zeitschrift fur Lebensmittel-Untersuchung und -Forschung, 189(1), 21-25 (1989-07-01)
This study describes a headspace gas-chromatographic method for the quantitative determination of inorganic bromide in vegetables. Different parameters influencing the reaction of the bromide ion into 2-bromoethanol have been investigated. The analysis of 2-bromoethanol is performed by the headspace--electron capture--gas
A R Jones et al.
Xenobiotica; the fate of foreign compounds in biological systems, 11(11), 763-770 (1981-11-01)
1. The metabolism of 2-bromo[U-14C]ethanol an [U-14C]ethylene oxide has been studied in the rat. 2. As both compounds give rise to similar amounts of two urinary metabolites, identified as S-(2-hydroxyethyl)cysteine and N-acetyl-S-(2-hydroxyethyl)cysteine, it is proposed that 2-bromoethanol is converted into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service