Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C45008

Sigma-Aldrich

6-Chlorohexanol

96%

Synonym(s):

1-Chloro-6-hydroxyhexane, Hexamethylene chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)6OH
CAS Number:
Molecular Weight:
136.62
Beilstein:
1697307
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

108-112 °C/14 mmHg (lit.)

density

1.024 g/mL at 25 °C (lit.)

SMILES string

OCCCCCCCl

InChI

1S/C6H13ClO/c7-5-3-1-2-4-6-8/h8H,1-6H2

InChI key

JNTPTNNCGDAGEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang Peng et al.
Journal of colloid and interface science, 531, 160-167 (2018-07-22)
The photocontrolled self-assembly of coumarin-functionalized silica nanoparticles is fabricated and investigated. When these nanoparticles are uniformly dispersed in CHCl3 solution under 365 nm light illumination, four novel kinds of patterns are self-assembled on the bottom of glass beakers: circle patterns, square
Ilaria Fregno et al.
The EMBO journal, 37(17) (2018-08-05)
Maintenance of cellular proteostasis relies on efficient clearance of defective gene products. For misfolded secretory proteins, this involves dislocation from the endoplasmic reticulum (ER) into the cytosol followed by proteasomal degradation. However, polypeptide aggregation prevents cytosolic dislocation and instead activates
Zhangting Xu et al.
Biomaterials science, 8(12), 3348-3358 (2020-05-28)
Hypoxia is an important pathological phenomenon due to uncontrolled cancer cell proliferation and insufficient blood flow, which can be used to design hypoxia-responsive nanocarriers for the intelligent treatment of tumor. However, it is difficult to obtain the response of hypoxia-responsive
Wen Sun et al.
Advanced healthcare materials, 5(4), 467-473 (2015-12-19)
The use of self-assembled nanostructures consisting of red-light-responsive Ru(II)-containing block copolymers (BCPs) for anticancer phototherapy is demonstrated. Three Ru-containing BCPs with different molecular weights are synthesized. Each BCP contains a hydrophilic poly(ethylene glycol) block and an Ru-containing block. In the
Leicheng Yin et al.
Soft matter, 11(30), 6145-6151 (2015-07-07)
A series of one-armed cholesterol-linked azobenzene molecules named CholXAzo with different spacers were synthesized, in which Chol6Azo was found to have induced blue phases (BPs) with a concentration of 4.0 wt%. Under irradiation of 385 nm UV light with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service