Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

667706

Sigma-Aldrich

2-(Bromomethyl)benzyl alcohol

95%

Synonym(s):

2-(Bromomethyl)benzenemethanol, 2-Hydroxymethylbenzylbromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9BrO
CAS Number:
Molecular Weight:
201.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

impurities

1-3% (2-((2-Bromomethyl)benzyloxy)methyl)phenyl)methanol
1-3% 1,2-Bis(bromomethyl)benzene

mp

66-68 °C

functional group

bromo
hydroxyl

storage temp.

2-8°C

SMILES string

OCc1ccccc1CBr

InChI

1S/C8H9BrO/c9-5-7-3-1-2-4-8(7)6-10/h1-4,10H,5-6H2

InChI key

JDBQNNKIOFSPOA-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service