Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B83401

Sigma-Aldrich

2-Methylbenzyl bromide

98%

Synonym(s):

α-Bromo-o-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2Br
CAS Number:
Molecular Weight:
185.06
Beilstein:
636333
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.575 (lit.)

bp

216-217 °C/742 mmHg (lit.)

mp

16-20 °C (lit.)

density

1.381 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1CBr

InChI

1S/C8H9Br/c1-7-4-2-3-5-8(7)6-9/h2-5H,6H2,1H3

InChI key

WGVYCXYGPNNUQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service