Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

658405

Sigma-Aldrich

(S)-4-(4-Aminobenzyl)-2(1H)-oxazolidinone

97%

Synonym(s):

(4S)-4-[(4-Aminophenyl)methyl]-2-oxazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O2
CAS Number:
Molecular Weight:
192.21
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

107-111 °C

SMILES string

Nc1ccc(C[C@H]2COC(=O)N2)cc1

InChI

1S/C10H12N2O2/c11-8-3-1-7(2-4-8)5-9-6-14-10(13)12-9/h1-4,9H,5-6,11H2,(H,12,13)/t9-/m0/s1

InChI key

WNAVSKJKDPLWBD-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-4-(4-Aminobenzyl)-2(1H)-oxazolidinone can be used as a starting material in the synthesis of:
  • Oxazolidinone derived 2-azetidinones as metal cation sensors.
  • Serotonin receptor agonist named zolmitriptan.
  • Schiff base derived zinc metal complexes of biological importance.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Convenient and Industrially Viable Process for Preparation of Zolmitriptan
Neelakandan K, et al.
Journal of Heterocyclic Chemistry, 51(S1), E332-E334 (2014)
D Manikandan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 239, 118484-118484 (2020-05-30)
The experimental geometry (XRD), vibrational (IR and Raman), electronic (UV-visible) and NMR spectra of (S)-4-(4-Amino-benzyl)-oxazolidin-2-one (ABO) have been corroborated with the corresponding first principle calculated values at DFT using hybrid B3LYP exchange correlation functional invoking 6-311++g(d, p) basis set. The
(S)-4-(4-aminobenzyl)-2-oxazolidinone based 2-azetidinones for antimicrobial application and luminescent sensing of divalent metal cations
Baruah S, et al.
Journal of Heterocyclic Chemistry, 57, 2498?2511-2498?2511 (2020)
Novel zinc (II) complexes of heterocyclic ligands as antimicrobial agents: synthesis, characterisation, and antimicrobial studies
Yamgar RS, et al.
Bioinorganic Chemistry and Applications, 2014 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service