Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

298883

Sigma-Aldrich

(S)-(−)-4-Isopropyl-2-oxazolidinone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
Beilstein:
3588198
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

optical activity

[α]20/D −18°, c = 6 in ethanol

optical purity

ee: 98% (GLC)

mp

70-73 °C (lit.)

SMILES string

CC(C)[C@H]1COC(=O)N1

InChI

1S/C6H11NO2/c1-4(2)5-3-9-6(8)7-5/h4-5H,3H2,1-2H3,(H,7,8)/t5-/m1/s1

InChI key

YBUPWRYTXGAWJX-RXMQYKEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent employed as a chiral ligand in dirhodium(II) complexes and as a chiral auxiliary in aldol addition reactions. Versatile chiral auxiliary for asymmetric synthesis. For a recent review, see Aldrichimica Acta.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 115, 9968-9968 (1993)
The Journal of Organic Chemistry, 58, 5107-5107 (1993)
Ager, D.J., et al.
Aldrichimica Acta, 30, 3-3 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service