Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

902640

Sigma-Aldrich

(S)-4-Methyl-2-oxazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO2
CAS Number:
Molecular Weight:
101.10
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Assay

≥95%

form

powder

mp

52-56 °C

storage temp.

2-8°C

SMILES string

C[C@H]1COC(=O)N1

InChI

1S/C4H7NO2/c1-3-2-7-4(6)5-3/h3H,2H2,1H3,(H,5,6)/t3-/m0/s1

InChI key

VAJFEOKPKHIPEN-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service