Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

642665

Sigma-Aldrich

6-Hydroxycoumarin

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O3
CAS Number:
Molecular Weight:
162.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

mp

249-253 °C (lit.)

functional group

ester

SMILES string

Oc1ccc2OC(=O)C=Cc2c1

InChI

1S/C9H6O3/c10-7-2-3-8-6(5-7)1-4-9(11)12-8/h1-5,10H

InChI key

CJIJXIFQYOPWTF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin-Lin Gan et al.
Acta pharmacologica Sinica, 38(12), 1683-1690 (2017-07-18)
α-Ketoglutarate (α-KG), a pivotal metabolite in energy metabolism, has been implicated in nonalcoholic fatty liver disease (NAFLD) and several cancers. It is recently proposed that plasma α-KG is a surrogate biomarker of NAFLD. Here, we report the development of a
Mohammed S Alqahtani et al.
Polymers, 12(11) (2020-11-06)
The main objective of the present study was to investigate the hemo and immune compatibility of gliadin nanoparticles as a function of particle size. Gliadin nanoparticles of different size were prepared using a modified antisolvent nanoprecipitation method. The hemolytic potential
Viviane C K N Deuschle et al.
Inflammopharmacology, 27(2), 323-338 (2018-01-13)
Persea americana, popularly known as avocado, has been empirically used as analgesic and anti-inflammatory including in the skin disorder treatment. Species of the genus Persea also show a photoprotective effect against UVB radiation. We investigated the antinociceptive and anti-inflammatory effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service