Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

417343

Sigma-Aldrich

6-Chloro-4-hydroxycoumarin

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5ClO3
CAS Number:
Molecular Weight:
196.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

291 °C (dec.) (lit.)

functional group

chloro
ester
ketone

SMILES string

OC1=CC(=O)Oc2ccc(Cl)cc12

InChI

1S/C9H5ClO3/c10-5-1-2-8-6(3-5)7(11)4-9(12)13-8/h1-4,11H

InChI key

HUMZENGQNOATEQ-UHFFFAOYSA-N

General description

6-Chloro-4-hydroxycoumarin is a substituted 4-hydroxycoumarin. It participates in the Fe/Acetic acid (AcOH) catalyzed synthesis of various biologically active compounds.

Application

6-Chloro-4-hydroxycoumarin may be employed for the synthesis of 5-alkylidene-4-chloro-5H-1,2,3-dithiazoles. It may be employed for the synthesis of 6-[(coumarinyl-1-aryl-propionyl]benzoxazinones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Iron/acetic acid mediated intermolecular tandem C-C and C-N bond formation: an easy access to acridinone and quinoline derivatives.
Rajawinslin RR, et al.
Royal Society of Chemistry Advances, 4(71), 37806-37811 (2014)
Da-Wei Yan et al.
Molecules (Basel, Switzerland), 25(2) (2020-01-16)
Natural meroterpenes derived from phloroglucinols and β-caryophyllene have shown high inhibitory activity against α-glucosidase or cancer cells, however, the chemical diversity of this type of skeletons in Nature is limited. To expand the chemical space and explore their inhibitory activities
Synthesis of new 5-alkylidene-4-chloro-5H-1, 2, 3-dithiazoles and their stereochemistry.
Jeon M-K and Kim K.
Tetrahedron, 55(32), 9651-9667 (1999)
Synthesis of 6-[1-(4'-hydroxycoumarin-3'-yl)-1-phenyl] propionyl-2H-[1,4]-benzoxazin-3(4H)-ones.
Jagath RG, et al.
Heterocyclic Communications, 10(1), 93-96 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service