Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

630233

Sigma-Aldrich

1-(Phenylsulfinyl)piperidine

97%

Synonym(s):

1-Benzenesulfinylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NOS
CAS Number:
Molecular Weight:
209.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

84-88 °C (lit.)

storage temp.

−20°C

SMILES string

O=S(N1CCCCC1)c2ccccc2

InChI

1S/C11H15NOS/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1,3-4,7-8H,2,5-6,9-10H2

InChI key

LBRJCAJLGAXDKP-UHFFFAOYSA-N

Application

Reagent for:
  • Selective glycosylation
  • Mannopyranolsylations
  • Design of enzyme stabilizers

Reagent for synthesis of:
  • Glycan array for affinity studies with influenza hemagglutinins†
  • Orthogonal protecting groups for preparation of highly branched oligosaccharides
  • Tetrasaccharide repeating units related to E. Coli 78 O-antigenic polysaccharides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Flavia Izzo et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(60), 15189-15193 (2017-08-24)
Unprotected tertiary sulfonimidamides have been prepared in good to excellent yields in a one-pot transformation from tertiary sulfinamides through NH transfer. The reaction is mediated by commercially available (diacetoxyiodo)benzene and ammonium carbamate in methanol under convenient conditions. A wide range

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service