Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

615951

Sigma-Aldrich

Palmitic acid-16,16,16-d3

≥99 atom % D, ≥99% (CP)

Synonym(s):

Hexadecanoic acid-16,16,16-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CH2)14CO2H
CAS Number:
Molecular Weight:
259.44
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % D

Quality Level

Assay

≥99% (CP)

form

solid

mp

61-64 °C

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CCCCCCCCCCCCCCC(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1D3

InChI key

IPCSVZSSVZVIGE-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pooranee K Morgan et al.
The Journal of biological chemistry, 297(6), 101341-101341 (2021-10-26)
Adipose-tissue-resident macrophages (ATMs) maintain metabolic homeostasis but also contribute to obesity-induced adipose tissue inflammation and metabolic dysfunction. Central to these contrasting effects of ATMs on metabolic homeostasis is the interaction of macrophages with fatty acids. Fatty acid levels are increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service