Skip to Content
MilliporeSigma
All Photos(1)

Documents

683582

Sigma-Aldrich

Oleic acid-d34

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)7CD=CD(CD2)7CO2D
CAS Number:
Molecular Weight:
316.67
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.13

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

liquid

bp

192-195 °C/1.2 mmHg (lit.)

mp

13.4 °C (lit.)

density

0.996 g/mL at 25 °C

mass shift

M+34

storage temp.

−20°C

SMILES string

[2H]OC(=O)C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(\[2H])=C(\[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9-/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D,10D,11D2,12D2,13D2,14D2,15D2,16D2,17D2/hD

InChI key

ZQPPMHVWECSIRJ-PRLROCHISA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qihong Zhang et al.
Pharmaceutical research, 31(10), 2762-2773 (2014-05-06)
To demonstrate the efficacy of infrared (IR) spectroscopic imaging for evaluation of lateral diffusion in stratum corneum (SC) and for elucidation of intermolecular interactions between exogenous agents and SC constituents. In separate experiments, acyl chain perdeuterated oleic acid (OA-d) and
Josephine Bradley et al.
Journal of cell science, 132(13) (2019-06-12)
Mammalian eggs generate most of their ATP by mitochondrial oxidation of pyruvate from the surrounding medium or from fatty acids that are stored as triacylglycerols within lipid droplets. The balance between pyruvate and fatty acid oxidation in generating ATP is
Chih-Chun Janet Lin et al.
Nature cell biology, 19(5), 550-557 (2017-04-25)
Microorganisms and their hosts share the same environment, and microbial metabolic molecules (metabolites) exert crucial effects on host physiology. Environmental factors not only shape the composition of the host's resident microorganisms, but also modulate their metabolism. However, the exact molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service