MilliporeSigma
  • An assembly and interaction of upconversion and plasmonic nanoparticles on organometallic nanofibers: enhanced multicolor upconversion, downshifting emission and the plasmonic effect.
An assembly and interaction of upconversion and plasmonic nanoparticles on organometallic nanofibers: enhanced multicolor upconversion, downshifting emission and the plasmonic effect.
Nanotechnology (2017-07-19)
Priyam Singh, Praveen Kumar Shahi, Rajiv Prakash, Shyam Bahadur Rai
ABSTRACT
We present novel inorganic-organic hybrid nanoparticles (HNPs) constituting inorganic NPs, NaY
Sigma-Aldrich
Terbium(III) oxide, 99.99% trace metals basis
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Erbium(III) oxide, ≥99.99% trace metals basis