Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

579688

Sigma-Aldrich

Lauric acid-2-13C

99 atom % 13C

Synonym(s):

Dodecanoic acid-2-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)913CH2CO2H
CAS Number:
Molecular Weight:
201.31
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99 atom % 13C

Quality Level

form

solid

bp

225 °C/100 mmHg (lit.)

mp

44-46 °C (lit.)

mass shift

M+1

SMILES string

CCCCCCCCCC[13CH2]C(O)=O

InChI

1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)/i11+1

InChI key

POULHZVOKOAJMA-KHWBWMQUSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F

Flash Point(C)

113.00 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service