Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616036

Sigma-Aldrich

N-Acetyl-DL-alanine-2-d

98 atom % D

Synonym(s):

DL-Alanine-2-d1, N-Acetyl derivative

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CD(NHCOCH3)CO2H
Molecular Weight:
132.14
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:

isotopic purity

98 atom % D

mass shift

M+1

SMILES string

[2H]C(C)(NC(C)=O)C(O)=O

InChI

1S/C5H9NO3/c1-3(5(8)9)6-4(2)7/h3H,1-2H3,(H,6,7)(H,8,9)/i3D

InChI key

KTHDTJVBEPMMGL-WFVSFCRTSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service