Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

485608

Sigma-Aldrich

Lauric acid-12,12,12-d3

98 atom % D

Synonym(s):

Dodecanoic acid-12,12,12-d3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CH2)10CO2H
CAS Number:
Molecular Weight:
203.34
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

99% (CP)

form

solid

bp

225 °C/100 mmHg (lit.)

mp

44-46 °C (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])CCCCCCCCCCC(O)=O

InChI

1S/C12H24O2/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3,(H,13,14)/i1D3

InChI key

POULHZVOKOAJMA-FIBGUPNXSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F

Flash Point(C)

113.00 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service