Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

570133

Sigma-Aldrich

Tris(dimethylamino)silane

electronic grade, 99.999%

Synonym(s):

(Me2N)3SiH, N,N,N′,N′,N′′,N′′-Hexamethylsilanetriamine, TDMAS, Tris(dimethylamido)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
((CH3)2N)3SiH
CAS Number:
Molecular Weight:
161.32
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

grade

electronic grade

description

570133

Assay

99.999%

reaction suitability

reagent type: reductant

bp

142 °C (lit.)

mp

−90 °C (lit.)

density

0.838 g/mL at 25 °C (lit.)

SMILES string

CN(C)[SiH](N(C)C)N(C)C

InChI

1S/C6H19N3Si/c1-7(2)10(8(3)4)9(5)6/h10H,1-6H3

InChI key

TWVSWDVJBJKDAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Please inquire for quantity, pricing, and packaging options.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

16.0 - 32.0 °F - closed cup

Flash Point(C)

-8.89 - 0.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B Ballantyne et al.
Toxicology and industrial health, 5(1), 45-54 (1989-01-01)
The acute handling hazards of tris(dimethylamino)silane [TDMAS] were investigated. The acute male rat peroral LD50 (with 95% confidence limits) was 0.71 (0.51-0.97) ml/kg, and the acute male rabbit percutaneous LD50 was 0.57 (0.35-0.92) ml/kg. The liquid was severely irritating to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service