Skip to Content
MilliporeSigma
Tris(dimethylamino)silane packaged for use in deposition systems

Tris(dimethylamino)silane

Tris(dimethylamino)silane packaged for use in deposition systems
Synonym(s):
(Me2N)3SiH, N,N,N′,N′,N′′,N′′-Hexamethylsilanetriamine, TDMAS, Tris(dimethylamido)silane, Tris(dimethylamino)silane
Linear Formula:
((CH3)2N)3SiH
CAS Number:
Molecular Weight:
161.32
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Showing 1-2 of 2