Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

550035

Sigma-Aldrich

2-(Dibutylamino)ethanol

99%

Synonym(s):

N,N-Dibutylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2NCH2CH2OH
CAS Number:
Molecular Weight:
173.30
Beilstein:
1738441
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6 (vs air)

Assay

99%

refractive index

n20/D 1.444 (lit.)

bp

229-230 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

CCCCN(CCO)CCCC

InChI

1S/C10H23NO/c1-3-5-7-11(9-10-12)8-6-4-2/h12H,3-10H2,1-2H3

InChI key

IWSZDQRGNFLMJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(Dibutylamino)ethanol is a tertiary amine.

Application

2-(Dibutylamino)ethanol (DBAE) may be used as a coreactant to enhance the electrochemiluminescence (ECL) of tris(2,2′-bipyridyl)ruthenium(II) [Ru(bpy)32+], employed in the ECL immunoassays and DNA probe assays. DBAE may also be used as a catalyst during the synthesis of protected D-erythro-(2S,3R)-sphingosine.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Environmentally Friendly and Highly Sensitive Ruthenium (II) Tris(2,2'-bipyridyl) Electrochemiluminescent System Using 2-(Dibutylamino) ethanol as Co-Reactant.
Liu X, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 119(3), 425-428 (2007)
Asymmetric alkenylation of chiral and prochiral aldehydes catalysed by chiral or achiral amino alcohols: catalytic diastereoselective synthesis of protected erythro-sphingosine and enantioselective synthesis of chiral diallyl alcohols.
Soai K and Takahashi K.
Journal of the Chemical Society. Perkin Transactions 1, 10, 1257-1258 (1994)
Zhiyong Guo et al.
Biosensors & bioelectronics, 95, 27-33 (2017-04-17)
Here, a novel potential-resolved "in-electrode" type electrochemiluminescence (ECL) immunosensor was fabricated based on two different types of luminant Ru-NH
Ting Bai et al.
Journal of colloid and interface science, 552, 439-447 (2019-06-01)
A drug delivery system that is integrated with fluorescent imaging is an emerging platform for tumor diagnostic and therapy. A pH-responsive fluorescent polymer that can respond to the surrounding medium is a desired component with which to construct an advanced
Stefanie Kirschbaum-Harriman et al.
The Analyst, 142(14), 2648-2653 (2017-06-15)
The influence of surfactants and coreactants on Ru(bpy)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service