Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

542873

Sigma-Aldrich

Pyrene-1-boronic acid

≥95.0%

Synonym(s):

1-Pyreneboronic acid, 1-Pyrenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11BO2
CAS Number:
Molecular Weight:
246.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

form

solid

mp

247-251 °C (lit.)

SMILES string

OB(O)c1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C16H11BO2/c18-17(19)14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9,18-19H

InChI key

MWEKPLLMFXIZOC-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed E El-Khouly et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 16(10), 1512-1518 (2017-08-02)
A light harvesting subphthalocyanine-pyrene dyad has been synthesized and characterized by linking pyrene (Py) with subphthalocyanine (SubPc) at its axial position with the B-O bond through the para position of the benzene group. Upon photoexcitation at the pyrene unit of
Hailei Zhang et al.
ACS applied materials & interfaces, 7(42), 23805-23811 (2015-10-10)
A novel fluorescence probe based on modified halloysite nanotubes (HNTs) by using 1-pyrenylboronic acid selectively grafted onto the inner surface of lumen was successfully achieved. The solid-state nuclear magnetic resonance ((13)C and (11)B), X-ray photoelectron spectroscopy (XPS) and Fourier transform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service