Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

527815

Sigma-Aldrich

4-Cyanophenylacetonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4CH2CN
CAS Number:
Molecular Weight:
142.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

100-104 °C (lit.)

SMILES string

N#CCc1ccc(cc1)C#N

InChI

1S/C9H6N2/c10-6-5-8-1-3-9(7-11)4-2-8/h1-4H,5H2

InChI key

QILKKAFYAFEWGU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of the solvent on the mode of coordination of 4-cyanophenylacetonitrile to silver (I) ion.
Boring WC and Iwamoto RT.
Inorgorganica Chimica Acta, 7, 264-266 (1973)
Tuning the nitrogen content and surface properties of nitrogen-doped carbon nanotubes synthesized using a nitrogen-containing ferrocenyl derivative and ethylbenzoate.
Ombaka LM, et al.
J. Mater. Sci., 50(3), 1187-1200 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service