Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

206539

Sigma-Aldrich

4-Ethynylbenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CC6H4CN
CAS Number:
Molecular Weight:
127.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

156-160 °C (lit.)

SMILES string

C#Cc1ccc(cc1)C#N

InChI

1S/C9H5N/c1-2-8-3-5-9(7-10)6-4-8/h1,3-6H

InChI key

LAGNMUUUMQJXBF-UHFFFAOYSA-N

General description

The water complexes formed with 4-ethynylbenzonitrile (4EBzN) were studied using IR-UV double resonance spectroscopy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surajit Maity et al.
The journal of physical chemistry. A, 114(32), 8337-8344 (2010-08-13)
The water complexes of 4-ethynylbenzonitrile (4EBzN) were investigated with IR-UV double resonance spectroscopy. Water interacts with the pi electron density of the C[triple bond]N group in 4EBzN, leading to the formation of a quasiplanar cyclic complex incorporating C-H...O and O-H...pi

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service