Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

521507

Sigma-Aldrich

4-Propylphenylboronic acid

≥95%

Synonym(s):

4-Propylbenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C6H4B(OH)2
CAS Number:
Molecular Weight:
164.01
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95%

mp

89-97 °C (lit.)

SMILES string

CCCc1ccc(cc1)B(O)O

InChI

1S/C9H13BO2/c1-2-3-8-4-6-9(7-5-8)10(11)12/h4-7,11-12H,2-3H2,1H3

InChI key

WLCGYIWOKVWFLB-UHFFFAOYSA-N

Application

Reactant for:
  • Palladium/carbon-catalyzed Suzuki coupling reactions
  • Preparation of biologically and pharmacologically active molecules

Other Notes

Contains varying amounts of anhydride

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandre Baralle et al.
Macromolecular rapid communications, 41(6), e1900644-e1900644 (2020-02-06)
In a significant breakthrough from classical molecular (i.e., nonpolymeric) iodonium salts in light-induced photochemistry, the synthesis and use of new safer polymeric iodonium salts are reported here. They are shown to be involved in charge transfer complexes (CTCs) while in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service