Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

499536

Sigma-Aldrich

4-Ethylphenylboronic acid

Synonym(s):

(ρ-Ethylphenyl)boronic acid, 4-Ethylbenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4B(OH)2
CAS Number:
Molecular Weight:
149.98
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

mp

150-155 °C (lit.)

SMILES string

CCc1ccc(cc1)B(O)O

InChI

1S/C8H11BO2/c1-2-7-3-5-8(6-4-7)9(10)11/h3-6,10-11H,2H2,1H3

InChI key

RZCPLOMUUCFPQA-UHFFFAOYSA-N

Application

Reactant for:
  • Suzuki-Miyaura cross-coupling reactions

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Daniel P O'Connell et al.
Bioorganic & medicinal chemistry letters, 22(3), 1397-1401 (2012-01-10)
Endothelial lipase (EL) and lipoprotein lipase (LPL) are homologous lipases that act on plasma lipoproteins. EL is predominantly a phospholipase and appears to be a key regulator of plasma HDL-C. LPL is mainly a triglyceride lipase regulating (V)LDL levels. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service