Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

646598

Sigma-Aldrich

Cyclobutylboronic acid

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9BO2
CAS Number:
Molecular Weight:
99.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

form

solid

mp

118-123 °C (lit.)

storage temp.

2-8°C

SMILES string

OB(O)C1CCC1

InChI

1S/C4H9BO2/c6-5(7)4-2-1-3-4/h4,6-7H,1-3H2

InChI key

MIUALDDWOKMYDA-UHFFFAOYSA-N

Application

  • Reagent used in palladium-catalyzed arylation and alkylation of diphenylisoxazole with boronic acids via C-H activated isoxazole palladacycle intermediate

  • Reagent used in the preparation of cyclobutyl arenes and heteroarenes via palladium catalyzed Suzuki-Miyaura cross-coupling reaction of potassium cyclobutyltrifluoroborate intermediate with aryl and heteroaryl chlorides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-Catalyzed Arylation and Alkylation of 3,5-Diphenylisoxazole with Boronic Acids via C-H Activation
J. Chu, et al.,
Organometallics, 27, 5173-5176 (2008)
Gary A Molander et al.
The Journal of organic chemistry, 73(19), 7481-7485 (2008-09-02)
Suitable conditions were found for the Suzuki-Miyaura cross-coupling reaction of potassium cyclopropyl- and cyclobutyltrifluoroborates with aryl chlorides. Both of these secondary alkyl trifluoroborates coupled in moderate to excellent yield with electron-rich, electron-poor, and hindered aryl chlorides to give a variety

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service