496863
Pentakis(dimethylamino)tantalum(V)
99.99%
Synonym(s):
PDMAT, TADMA, Ta(NMe2)5, Tantalum dimethylamide
About This Item
Recommended Products
Assay
99.99%
form
solid
reaction suitability
core: tantalum
reagent type: catalyst
mp
100 °C (dec.) (lit.)
SMILES string
CN(C)[Ta](N(C)C)(N(C)C)(N(C)C)N(C)C
InChI
1S/5C2H6N.Ta/c5*1-3-2;/h5*1-2H3;/q5*-1;+5
InChI key
VSLPMIMVDUOYFW-UHFFFAOYSA-N
General description
Features and Benefits
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 1
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Thin film photovoltaic devices have become increasingly important in efficiently harnessing solar energy to meet consumer demand.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service