Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

218634

Sigma-Aldrich

Tantalum(V) chloride

99.8% trace metals basis

Synonym(s):

Tantalum pentachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TaCl5
CAS Number:
Molecular Weight:
358.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

99.8% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: tantalum

impurities

≤2500.0 ppm Trace Metal Analysis

density

3.68 g/mL at 25 °C (lit.)

SMILES string

Cl[Ta](Cl)(Cl)(Cl)Cl

InChI

1S/5ClH.Ta/h5*1H;/q;;;;;+5/p-5

InChI key

OEIMLTQPLAGXMX-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Application

Used to form mixed chloro-aryloxide compounds, such as [Ta(OC6H3-2,6-i-Pr2)2Cl3]2, which form pyridine or phosphine adducts with a distorted six-coordinate geometry. Also used as a starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18, for which electron and stability studies were performed.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janet R. Clark et al.
Inorganic chemistry, 36(17), 3623-3631 (1997-08-13)
The mixed chloro-aryloxide compounds [M(OC(6)H(3)Pr(i)(2)-2,6)(2)Cl(3)](2) (M = Nb (1a), Ta (1b); both structurally characterized) and [M(OC(6)H(3)Pr(i)(2)-2,6)(3)Cl(2)] (M = Nb (2a), Ta (2b)) react with pyridine (py) and PMe(2)Ph to produce a series of adducts cis-mer-[MCl(3)(OC(6)H(3)Pr(i)(2)-2,6)(2)(L)] (M, L: Nb, py (3a);
Ogliaro, F. et al.
Inorganic Chemistry, 37, 6199-6199 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service