Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

487228

Sigma-Aldrich

1,3-Butadiene-d6

≥98 atom % D, ≥98% (CP), contains hydroquinone as stabilizer

Synonym(s):

Perdeutero-1,3-butadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
D2C=CDCD=CD2
CAS Number:
Molecular Weight:
60.13
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

vapor pressure

3102 mmHg ( 37.7 °C)

Quality Level

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

autoignition temp.

788 °F

contains

hydroquinone as stabilizer

bp

−4.5 °C (lit.)

mp

−109 °C (lit.)

mass shift

M+6

SMILES string

[2H]\C([2H])=C([2H])\C([2H])=C(\[2H])[2H]

InChI

1S/C4H6/c1-3-4-2/h3-4H,1-2H2/i1D2,2D2,3D,4D

InChI key

KAKZBPTYRLMSJV-UFSLNRCZSA-N

Related Categories

Packaging

1 L quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressure at 21ºC is 20 psig for 1 L. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951 or Z187224
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Flam. Gas 1A - Muta. 1B - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service