Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

486701

Sigma-Aldrich

Boc-Gly-OH-15N

98 atom % 15N, 99% (CP)

Synonym(s):

N-(tert-Butoxycarbonyl)glycine-15N, 15N Labeled Boc-Gly-OH, 15N Labeled N-(tert-butoxycarbonyl)glycine, Glycine-15N, N-t-Boc derivative

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Boc-15NHCH2CO2H
CAS Number:
Molecular Weight:
176.18
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

Assay

99% (CP)

form

solid

technique(s)

bio NMR: suitable

mp

86-89 °C (lit.)

application(s)

peptide synthesis

functional group

Boc

mass shift

M+1

SMILES string

CC(C)(C)OC(=O)[15NH]CC(O)=O

InChI

1S/C7H13NO4/c1-7(2,3)12-6(11)8-4-5(9)10/h4H2,1-3H3,(H,8,11)(H,9,10)/i8+1

InChI key

VRPJIFMKZZEXLR-VJJZLTLGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service