Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

484903

Sigma-Aldrich

Piperazine-2-carboxylic acid dihydrochloride

98%

Synonym(s):

(±)-Piperazine-2-carboxylic acid dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O2 · 2HCl
CAS Number:
Molecular Weight:
203.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

265 °C (dec.) (lit.)

SMILES string

Cl[H].Cl[H].OC(=O)C1CNCCN1

InChI

1S/C5H10N2O2.2ClH/c8-5(9)4-3-6-1-2-7-4;;/h4,6-7H,1-3H2,(H,8,9);2*1H

InChI key

WNSDZBQLMGKPQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maryam Hashemi et al.
Materials science & engineering. C, Materials for biological applications, 61, 791-800 (2016-02-04)
Poly-(amidoamine) (PAMAM) and poly-(propylenimine) (PPI) are the two most widely investigated dendrimers for drug and gene delivery. In order to enhance DNA transfection activity of these dendrimers, generation 3 and 4 PAMAM and generation 4 and 5 PPI were modified
Zahra Salmasi et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 96, 76-88 (2015-07-26)
Branched polyethylenimine (PEI) is extensively used as a polycationic non-viral vector for gene delivery. Polyplexes formed with PEI are believed to be released from endocytotic vesicles by the osmotic burst mechanism in the rate-limiting step in transfection. Increasing the buffering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service