Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

484237

Sigma-Aldrich

Di(propylene glycol) butyl ether, mixture of isomers

≥98.5%

Synonym(s):

DOWANOL® DPnB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OC3H6OC3H6OH
CAS Number:
Molecular Weight:
190.28
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Assay

≥98.5%

refractive index

n20/D 1.426 (lit.)

bp

222-232 °C (lit.)

density

0.913 g/mL at 25 °C (lit.)

SMILES string

O(C(COCCCC)C)CC(O)C

InChI

1S/C10H22O3/c1-4-5-6-12-8-10(3)13-7-9(2)11/h9-11H,4-8H2,1-3H3

InChI key

CUVLMZNMSPJDON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Dow Chemical
DOWANOL is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service