Skip to Content
MilliporeSigma
All Photos(1)

Documents

449393

Sigma-Aldrich

Di(ethylene glycol) hexyl ether

95%

Synonym(s):

2-(2-Hexyloxyethoxy)ethanol, C6E2, Hexyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
190.28
Beilstein:
1743959
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

refractive index

n20/D 1.4381 (lit.)

bp

260 °C (lit.)

mp

−40 °C (lit.)

density

0.935 g/mL at 25 °C (lit.)

SMILES string

CCCCCCOCCOCCO

InChI

1S/C10H22O3/c1-2-3-4-5-7-12-9-10-13-8-6-11/h11H,2-10H2,1H3

InChI key

GZMAAYIALGURDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

253.4 °F

Flash Point(C)

123 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Ballantyne et al.
Journal of applied toxicology : JAT, 21(6), 449-460 (2001-12-18)
Diethylene glycol monohexyl ether (DEGHE; CAS no. 112-59-4), an industrial chemical, was investigated for the potential to produce genotoxic effects using three in vitro and two in vivo tests. No mutagenic activity occurred in either the absence or presence of
B Ballantyne et al.
Veterinary and human toxicology, 29(5), 361-366 (1987-10-01)
Ethylene glycol monohexyl ether (EGHE) and diethylene glycol monohexyl ether (DGHE) are glycol ethers used as industrial solvents and coating materials, and whose acute handling hazards were investigated. Acute peroral LD50 values in the rat were for EGHE 1.67 ml/kg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service