Skip to Content
MilliporeSigma
All Photos(1)

Documents

479535

Sigma-Aldrich

Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution

0.05 M in poly(dimethylsiloxane), vinyl terminated

Synonym(s):

Karstedt′s catalyst, Platinum(0) 1,3-diethenyl-1,1,3,3-tetramethyldisiloxane complexes

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O[Si(CH3)2CH=CH2]2Pt
CAS Number:
Molecular Weight:
381.48
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: platinum
reagent type: catalyst

Quality Level

concentration

0.05 M in poly(dimethylsiloxane), vinyl terminated

bp

>200 °C

density

0.984 g/mL at 25 °C

SMILES string

[Pt].C[Si](C)(O[Si](C)(C)C=C)C=C

InChI

1S/C8H18OSi2.Pt/c1-7-10(3,4)9-11(5,6)8-2;/h7-8H,1-2H2,3-6H3;

InChI key

RCNRJBWHLARWRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service