Skip to Content
MilliporeSigma
All Photos(1)

Documents

482196

Sigma-Aldrich

Poly(dimethylsiloxane-co-methylhydrosiloxane), trimethylsilyl terminated

average Mn ~950, methylhydrosiloxane 50 mol %

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

average Mn ~950

composition

methylhydrosiloxane, 50 mol %

refractive index

n20/D 1.401

viscosity

12 cSt(lit.)

bp

>93 °C (lit.)

density

0.99 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.1 °F - closed cup

Flash Point(C)

65.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

See All

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service