Skip to Content
MilliporeSigma
Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution 0.05 M in poly(dimethylsiloxane), vinyl terminated

Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution

Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution 0.05 M in poly(dimethylsiloxane), vinyl terminated
Linear Formula:
O[Si(CH3)2CH=CH2]2Pt
CAS Number:
Molecular Weight:
381.48

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Chemical Composition
Markush Class
Markush Group
Reagent Type
Core