Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

477842

Sigma-Aldrich

PAMAM-OH dendrimer, generation 3 solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=3);dendri PAMAM(NHCH2CH2OH)32
Molecular Weight:
6940.35
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

amidoethanol surface groups

form

liquid

mol wt

generation 3

feature

core type ethylenediamine core (2-carbon core)

concentration

20 wt. % in methanol

no. Surface Groups

32

refractive index

n20/D 1.364

bp

65 °C

density

0.86 g/mL at 25 °C

storage temp.

2-8°C

Physical form

Contains 32 surface hydroxyl groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service