Skip to Content
MilliporeSigma

09193

4-Amino-L-phenylalanine hydrochloride

≥96.0% (calc. on dry substance, AT)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H12N2O2 · HCl
CAS Number:
Molecular Weight:
216.66
UNSPSC Code:
12352209
NACRES:
NA.22
PubChem Substance ID:
EC Number:
263-388-2
Beilstein/REAXYS Number:
3657715
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥96.0% (calc. on dry substance, AT)

form

powder

optical activity

[α]20/D +8.7±0.5°, c = 1% in acetic acid: water (4:1)

reaction suitability

reaction type: solution phase peptide synthesis

loss

≤5.0% loss on drying

color

white to faint beige

application(s)

peptide synthesis

SMILES string

Cl.N[C@@H](Cc1ccc(N)cc1)C(O)=O

InChI

1S/C9H12N2O2.ClH/c10-7-3-1-6(2-4-7)5-8(11)9(12)13;/h1-4,8H,5,10-11H2,(H,12,13);1H/t8-;/m0./s1

InChI key

ISCZSPZLBJLJGO-QRPNPIFTSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library