Skip to Content
MilliporeSigma

B1375

Bathophenanthrolinedisulfonic acid disodium salt hydrate

≥95%

Synonym(s):

4,7-Diphenyl-1,10-phenanthroline disulfonic acid disodium salt hydrate, 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H14N2Na2O6S2 · xH2O
CAS Number:
Molecular Weight:
536.49 (anhydrous basis)
MDL number:
NACRES:
NA.47
UNSPSC Code:
12171500
Assay:
≥95%
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95%

form

powder

mp

300 °C (lit.)

solubility

H2O: 100 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Application

Bathophenanthrolinedisulfonic acid disodium salt hydrate has been used to measure iron to determine the solubility of iron in the biological sample. It has been used as an iron chelator to grow plants without Fe-EDTA (ethylenediaminetetraacetic acid).

Biochem/physiol Actions

Bathophenanthrolinedisulfonic acid is a metal chelator. It permits iron determination to be made in aqueous medium without need for solvent extraction. It forms a complex with Fe2+ and makes it unavailable to Fe2+ transporter, thereby inhibiting the transport of iron into chloroplasts. Bathophenanthrolinedisulfonic acid is also used in the assay for determining intracellular glutathione.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library