Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47628

Sigma-Aldrich

Fmoc-Ile-OH

≥98.0% (T)

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-isoleucine, Fmoc-L-isoleucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
Beilstein:
4716717
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98.0% (T)

form

solid

optical activity

[α]20/D −12±1°, c = 1% in DMF

reaction suitability

reaction type: C-H Activation
reaction type: Fmoc solid-phase peptide synthesis
reagent type: ligand
reaction type: Peptide Synthesis

mp

145-147 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc
amine
carboxylic acid

storage temp.

2-8°C

SMILES string

CC[C@H](C)[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C21H23NO4/c1-3-13(2)19(20(23)24)22-21(25)26-12-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,13,18-19H,3,12H2,1-2H3,(H,22,25)(H,23,24)/t13-,19-/m0/s1

InChI key

QXVFEIPAZSXRGM-DJJJIMSYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moise Ndao et al.
Langmuir : the ACS journal of surfaces and colloids, 34(29), 8678-8684 (2016-04-05)
Diatoms are unicellular algae that construct cell walls called frustules by the precipitation of silica, using special proteins that order the silica into a wide variety of nanostructures. The diatom species Cylindrotheca fusiformis contains proteins called silaffins within its frustules
Karthik Nadendla et al.
Molecular pharmaceutics, 16(7), 2922-2928 (2019-05-24)
We have previously described the photoactivated depot (PAD) approach for the light-stimulated release of therapeutic proteins such as insulin. The aim of this method is to release insulin from a shallow dermal depot in response to blood glucose information, using
Yu-I Hsu et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 108(4), 1274-1280 (2019-08-21)
We recently reported a novel method to modify the polylactic acid (PLLA) nanofiber scaffold with IKVAV peptide/oligo (d-lactic acid) (ODLA) heteroconjugates. However, a part of the IKVAV sequence was not exposed to the outermost surface and therefore, cell adhesion was
R Ashton Lavoie et al.
International journal of molecular sciences, 20(7) (2019-04-11)
The growing integration of quality-by-design (QbD) concepts in biomanufacturing calls for a detailed and quantitative knowledge of the profile of impurities and their impact on the product safety and efficacy. Particularly valuable is the determination of the residual level of
R Ashton Lavoie et al.
Biotechnology and bioengineering, 117(2), 438-452 (2019-10-28)
The clearance of host cell proteins (HCPs) is of crucial importance in biomanufacturing, given their diversity in composition, structure, abundance, and occasional structural homology with the product. The current approach to HCP clearance in the manufacturing of monoclonal antibodies (mAbs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service