Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

47623

Sigma-Aldrich

Fmoc-Tyr(tBu)-OH

≥98.0% (HPLC), for peptide synthesis

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-tyrosine, Fmoc-O-tert-butyl-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H29NO5
CAS Number:
Molecular Weight:
459.53
Beilstein:
4216652
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

Fmoc-Tyr(tBu)-OH, ≥98.0% (HPLC)

Assay

≥98.0% (HPLC)

form

powder

optical activity

[α]20/D −29±2°, c = 1% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

153-156 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)Oc1ccc(C[C@H](NC(=O)OCC2c3ccccc3-c4ccccc24)C(O)=O)cc1

InChI

1S/C28H29NO5/c1-28(2,3)34-19-14-12-18(13-15-19)16-25(26(30)31)29-27(32)33-17-24-22-10-6-4-8-20(22)21-9-5-7-11-23(21)24/h4-15,24-25H,16-17H2,1-3H3,(H,29,32)(H,30,31)/t25-/m0/s1

InChI key

JAUKCFULLJFBFN-VWLOTQADSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fmoc-Tyr(tBu)-OH also known as Fmoc-O-tert-butyl-L-tyrosine, is commonly used in Fmoc solid phase peptide synthesis.

Application

Fmoc-Tyr(tBu)-OH can be used to synthesize Leu-EnkephalinAmide via solid phase synthesis in water. Additionally, it can be used to synthesize amino acid derivatives such as Fmoc-Tyr-OAllyl.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-Phase Total Synthesis of the Proposed Structure of Coibamide A and Its Derivative: Highly Methylated Cyclic Depsipeptides
GA Sable
European Journal of Organic Chemistry, 2015, 7043-7052 (2015)
Solid-phase peptide synthesis using nanoparticulate amino acids in water
K Hojo
Journal of Peptide Science, 13, 493-497 (2007)
Rini Roy et al.
Journal of the American Society for Mass Spectrometry, 29(6), 1086-1098 (2018-05-26)
Immunoglobulins, such as immunoglobulin G (IgG), are of prime importance in the immune system. Polyclonal human IgG comprises four subclasses, of which IgG1 and IgG2 are the most abundant in healthy individuals. In an effort to develop an absolute MALDI-ToF-MS
Chloé Guilbaud-Chéreau et al.
ACS applied materials & interfaces, 11(14), 13147-13157 (2019-03-14)
Molecular gels formed by the self-assembly of low-molecular-weight gelators have received increasing interest because of their potential applications in drug delivery. In particular, the ability of peptides and amino acids to spontaneously self-assemble into three-dimensional fibrous network has been exploited
Loay Awad et al.
Chembiochem : a European journal of chemical biology, 17(24), 2353-2360 (2016-10-28)
We present the design, synthesis, and characterization of a novel photocaged glutamine derivative (modified on the side chain of glutamine), and describe its use in enhancing peptide stability and solubility. Our results demonstrate that this approach can be used to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service