Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

466085

Sigma-Aldrich

Cerium(III) iodide

AnhydroBeads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Cerous triiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeI3
CAS Number:
Molecular Weight:
520.83
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for synthesis
synthesis grade

grade

for synthesis
synthesis grade

product line

AnhydroBeads

Assay

99.99% trace metals basis

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤150.0 ppm Trace Rare Earth Analysis

particle size

−10 mesh

mp

750 °C (lit.)

SMILES string

I[Ce](I)I

InChI

1S/Ce.3HI/h;3*1H/q+3;;;/p-3

InChI key

ZEDZJUDTPVFRNB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service