Skip to Content
MilliporeSigma
All Photos(1)

Documents

466336

Sigma-Aldrich

Strontium iodide

anhydrous, ≥99.99% trace metals basis

Synonym(s):

Hydriodic acid strontium salt (2:1), Strontium diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrI2
CAS Number:
Molecular Weight:
341.43
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Assay

≥99.99% trace metals basis

form

solid

impurities

≤100.0 ppm Trace Metal Analysis

bp

1773 °C/1 atm (lit.)

mp

515 °C (lit.)

density

4.55 g/mL at 25 °C (lit.)

SMILES string

I[Sr]I

InChI

1S/2HI.Sr/h2*1H;/q;;+2/p-2

InChI key

KRIJWFBRWPCESA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Strontium iodide is an ionic deliquescent compound with relatively high density, optical clarity, and high scintillation light yield. It is widely used in the field of perovskite solar cells and gamma radiation detectors.

Application

Strontium iodide can be used as a dopant in methylammonium lead iodide perovskite films. The addition of Sr2+ increases the charge carrier collection efficiency of solar cells resulting in very high fill factors.

It can be used as a precursor to prepare high-light yield scintillators with excellent X-ray and gamma-ray detection properties.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strontium insertion in methylammonium lead iodide: long charge carrier lifetime and high fill-factor solar cells
Daniel Perez-del-Rey, et al.
Advanced Materials, 28, 9839-9845 (2016)
Growth and characterization of potassium strontium iodide: A new high light yield scintillator with 2.4% energy resolution
L. Stand, et al.},
Nuclear Instruments and Methods in Physics Research, 780, 40-44 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service